RIE156330028 Rieke Metals, Inc.

5-(4-Chloro-2-methoxyphenyl)-5-oxovaleric acid

  • MDL number MFCD09801749
  • Molecular Weight : 256.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7204C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7204C
Availability details for
Quantity
  • Availability
  • Add to Cart