RIE156330030 Rieke Metals, Inc.

7-(4-Chloro-2-methoxyphenyl)-7-oxoheptanoic acid

  • MDL number MFCD09801751
  • Molecular Weight : 284.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7204E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7204E
Availability details for
Quantity
  • Availability
  • Add to Cart