RIE156330031 Rieke Metals, Inc.

8-(4-Chloro-2-methoxyphenyl)-8-oxooctanoic acid

  • MDL number MFCD09801752
  • Molecular Weight : 298.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7204F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7204F
Availability details for
Quantity
  • Availability
  • Add to Cart