RIE156330032 Rieke Metals, Inc.

6-(5-Chloro-2-methoxyphenyl)-6-oxohexanoic acid

  • MDL number MFCD09801754
  • Cas Number : 854879-20-2
  • Molecular Weight : 270.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7205D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7205D
Availability details for
Quantity
  • Availability
  • Add to Cart