RIE156330035 Rieke Metals, Inc.

4-[2-Chloro-4-(methylthio)phenyl]-4-oxobutyric acid

  • MDL number MFCD09801757
  • Cas Number : 103860-95-3
  • Molecular Weight : 258.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7206B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7206B
Availability details for
Quantity
  • Availability
  • Add to Cart