RIE156330036 Rieke Metals, Inc.

5-(2-Chloro-4-(methylthio)phenyl)-5-oxovaleric acid

  • MDL number MFCD09801758
  • Molecular Weight : 272.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7206C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7206C
Availability details for
Quantity
  • Availability
  • Add to Cart