RIE156330038 Rieke Metals, Inc.

7-[2-Chloro-4-(methylthio)phenyl]-7-oxoheptanoic acid

  • MDL number MFCD09801760
  • Molecular Weight : 300.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7206E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7206E
Availability details for
Quantity
  • Availability
  • Add to Cart