RIE156330039 Rieke Metals, Inc.

8-[2-Chloro-4-(methylthio)phenyl]-8-oxooctanoic acid

  • MDL number MFCD09801761
  • Molecular Weight : 314.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7206F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7206F
Availability details for
Quantity
  • Availability
  • Add to Cart