RIE156330040 Rieke Metals, Inc.

4-(2-Chloro-6-fluorophenyl)-4-oxobutyric acid

  • MDL number MFCD09801762
  • Molecular Weight : 230.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7207B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7207B
Availability details for
Quantity
  • Availability
  • Add to Cart