RIE156330045 Rieke Metals, Inc.

4-(2-Chloro-4,5-difluorophenyl)-4-oxobutyric acid

  • MDL number MFCD09801767
  • Molecular Weight : 248.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7208B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7208B
Availability details for
Quantity
  • Availability
  • Add to Cart