RIE156330046 Rieke Metals, Inc.

5-(2-Chloro-4,5-difluorophenyl)-5-oxovaleric acid

  • MDL number MFCD09801768
  • Molecular Weight : 262.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7208C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7208C
Availability details for
Quantity
  • Availability
  • Add to Cart