RIE156330048 Rieke Metals, Inc.

7-(2-Chloro-4,5-difluorophenyl)-7-oxoheptanoic acid

  • MDL number MFCD09801770
  • Molecular Weight : 290.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7208E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7208E
Availability details for
Quantity
  • Availability
  • Add to Cart