RIE156330050 Rieke Metals, Inc.

4-(2,3,4,5-tetrafluorophenyl)-4-oxobutyric acid

  • MDL number MFCD02261134
  • Molecular Weight : 250.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7209B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7209B
Availability details for
Quantity
  • Availability
  • Add to Cart