RIE156330051 Rieke Metals, Inc.

5-(2,3,4,5-tetrafluorophenyl)-5-oxovaleric acid

  • MDL number MFCD02261135
  • Molecular Weight : 264.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7209C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7209C
Availability details for
Quantity
  • Availability
  • Add to Cart