RIE156330052 Rieke Metals, Inc.

6-(2,3,4,5-tetrafluorophenyl)-6-oxohexanoic acid

  • MDL number MFCD02261136
  • Molecular Weight : 278.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7209D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7209D
Availability details for
Quantity
  • Availability
  • Add to Cart