RIE156330054 Rieke Metals, Inc.

8-(2,3,4,5-tetrafluorophenyl)-8-oxooctanoic acid

  • MDL number MFCD02261138
  • Molecular Weight : 306.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7209F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7209F
Availability details for
Quantity
  • Availability
  • Add to Cart