RIE156330055 Rieke Metals, Inc.

4-(3,4,5-trichlorophenyl)-4-oxobutyric acid

  • MDL number MFCD09801772
  • Cas Number : 62903-19-9
  • Molecular Weight : 281.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7210B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7210B
Availability details for
Quantity
  • Availability
  • Add to Cart