RIE156330057 Rieke Metals, Inc.

6-(3,4,5-trichlorophenyl)-6-oxohexanoic acid

  • MDL number MFCD09801774
  • Molecular Weight : 309.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7210D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7210D
Availability details for
Quantity
  • Availability
  • Add to Cart