RIE156330059 Rieke Metals, Inc.

8-(3,4,5-trichlorophenyl)-8-oxooctanoic acid

  • MDL number MFCD09801776
  • Molecular Weight : 337.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7210F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7210F
Availability details for
Quantity
  • Availability
  • Add to Cart