RIE156330065 Rieke Metals, Inc.

7-(2,4,5-trimethoxyphenyl)-7-oxoheptanoic acid

  • MDL number MFCD09801782
  • Molecular Weight : 310.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7212E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7212E
Availability details for
Quantity
  • Availability
  • Add to Cart