RIE156330067 Rieke Metals, Inc.

5-(2,3,4-trimethoxyphenyl)-5-oxovaleric acid

  • MDL number MFCD09801785
  • Cas Number : 16093-16-6
  • Molecular Weight : 282.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7213C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7213C
Availability details for
Quantity
  • Availability
  • Add to Cart