RIE156330068 Rieke Metals, Inc.

6-(2,3,4-trimethoxyphenyl)-6-oxohexanoic acid

  • MDL number MFCD09801786
  • Cas Number : 917591-97-0
  • Molecular Weight : 296.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7213D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7213D
Availability details for
Quantity
  • Availability
  • Add to Cart