RIE156330070 Rieke Metals, Inc.

8-(2,3,4-trimethoxyphenyl)-8-oxooctanoic acid

  • MDL number MFCD09801788
  • Molecular Weight : 324.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7213F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7213F
Availability details for
Quantity
  • Availability
  • Add to Cart