RIE156330071 Rieke Metals, Inc.

6-(4-n-Butylphenyl)-6-oxohexanoic acid

  • MDL number MFCD02260958
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7214D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7214D
Availability details for
Quantity
  • Availability
  • Add to Cart