RIE156330072 Rieke Metals, Inc.

7-(4-n-Butylphenyl)-7-oxoheptanoic acid

  • MDL number MFCD02260959
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7214E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7214E
Availability details for
Quantity
  • Availability
  • Add to Cart