RIE156330073 Rieke Metals, Inc.

8-(4-n-Butylphenyl)-8-oxooctanoic acid

  • MDL number MFCD02260960
  • Molecular Weight : 290.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7214F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7214F
Availability details for
Quantity
  • Availability
  • Add to Cart