RIE156330077 Rieke Metals, Inc.

4-[2-(Methylthio)phenyl]-4-oxobutyric acid

  • MDL number MFCD09801789
  • Molecular Weight : 224.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7216B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7216B
Availability details for
Quantity
  • Availability
  • Add to Cart