RIE156330078 Rieke Metals, Inc.

5-(2-(Methylthio)phenyl)-5-oxovaleric acid

  • MDL number MFCD09801790
  • Cas Number : 930116-83-9
  • Molecular Weight : 238.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7216C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7216C
Availability details for
Quantity
  • Availability
  • Add to Cart