RIE156330079 Rieke Metals, Inc.

4-(3-Ethoxyphenyl)-4-oxobutyric acid

  • MDL number MFCD09801791
  • Cas Number : 905592-33-8
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7218B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7218B
Availability details for
Quantity
  • Availability
  • Add to Cart