RIE156330082 Rieke Metals, Inc.

5-(3-Methyl-2-thienyl)-5-oxovaleric acid

  • MDL number MFCD09801794
  • Molecular Weight : 212.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7219C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7219C
Availability details for
Quantity
  • Availability
  • Add to Cart