RIE156330085 Rieke Metals, Inc.

4-(4-tert-Butyl-2,6-dimethylphenyl)-4-oxobutyric acid

  • MDL number MFCD09801795
  • Molecular Weight : 262.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7224B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7224B
Availability details for
Quantity
  • Availability
  • Add to Cart