RIE156330086 Rieke Metals, Inc.

5-(4-tert-Butyl-2,6-dimethylphenyl)-5-oxovaleric acid

  • MDL number MFCD09801796
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7224C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7224C
Availability details for
Quantity
  • Availability
  • Add to Cart