RIE156330089 Rieke Metals, Inc.

4-(3-Fluoro-2-methylphenyl)-4-oxobutyric acid

  • MDL number MFCD09801801
  • Molecular Weight : 210.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7229B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7229B
Availability details for
Quantity
  • Availability
  • Add to Cart