RIE156330094 Rieke Metals, Inc.

5-(5-Fluoro-2-methoxyphenyl)-5-oxovaleric acid

  • MDL number MFCD09801806
  • Molecular Weight : 240.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7232C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7232C
Availability details for
Quantity
  • Availability
  • Add to Cart