RIE156330095 Rieke Metals, Inc.

4-(3-Chloro-4-methylphenyl)-4-oxobutyric acid

  • MDL number MFCD09801809
  • Cas Number : 62903-06-4
  • Molecular Weight : 226.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7240B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7240B
Availability details for
Quantity
  • Availability
  • Add to Cart