RIE156330098 Rieke Metals, Inc.

3-Methyl-5-(2-methylphenyl)-5-oxovaleric acid

  • MDL number MFCD09801812
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7242

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7242
Availability details for
Quantity
  • Availability
  • Add to Cart