RIE156330099 Rieke Metals, Inc.

2,2-Dimethyl-4-(3-methylphenyl)-4-oxobutyric acid

  • MDL number MFCD09801813
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7243

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7243
Availability details for
Quantity
  • Availability
  • Add to Cart