RIE156330100 Rieke Metals, Inc.

2,2-Dimethyl-4-(4-methylphenyl)-4-oxobutyric acid

  • MDL number MFCD09801814
  • Cas Number : 71821-98-2
  • Molecular Weight : 220.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7245

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7245
Availability details for
Quantity
  • Availability
  • Add to Cart