RIE156330101 Rieke Metals, Inc.

2,2-Dimethyl-4-(1-naphthyl)-4-oxobutyric acid

  • MDL number MFCD09801815
  • Cas Number : 854904-16-8
  • Molecular Weight : 256.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7247

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7247
Availability details for
Quantity
  • Availability
  • Add to Cart