RIE156330102 Rieke Metals, Inc.

3-Methyl-5-(1-naphthyl)-5-oxovaleric acid

  • MDL number MFCD09801816
  • Molecular Weight : 256.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7248

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7248
Availability details for
Quantity
  • Availability
  • Add to Cart