RIE156330103 Rieke Metals, Inc.

2,2-Dimethyl-4-[2-(methylthio)phenyl]-4-oxobutyric acid

  • MDL number MFCD09801817
  • Molecular Weight : 252.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7249

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7249
Availability details for
Quantity
  • Availability
  • Add to Cart