RIE156330111 Rieke Metals, Inc.

2,2-Dimethyl-4-[2-(ethylthio)phenyl]-4-oxobutyric acid

  • MDL number MFCD09801825
  • Molecular Weight : 266.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7257

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7257
Availability details for
Quantity
  • Availability
  • Add to Cart