RIE156330115 Rieke Metals, Inc.

2,2-Dimethyl-4-oxo-4-(4-phenoxyphenyl)butyric acid

  • MDL number MFCD09801829
  • Molecular Weight : 298.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7261

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7261
Availability details for
Quantity
  • Availability
  • Add to Cart