RIE156330116 Rieke Metals, Inc.

3-Methyl-5-oxo-5-(4-phenoxyphenyl)valeric acid

  • MDL number MFCD09801830
  • Cas Number : 64157-56-8
  • Molecular Weight : 298.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7262

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7262
Availability details for
Quantity
  • Availability
  • Add to Cart