RIE156330118 Rieke Metals, Inc.

5-(4-Isopropylphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801832
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7264

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7264
Availability details for
Quantity
  • Availability
  • Add to Cart