RIE156330123 Rieke Metals, Inc.

2,2-Dimethyl-4-oxo-4-(4-n-propylphenyl)butyric acid

  • MDL number MFCD09801837
  • Molecular Weight : 248.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7271

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7271
Availability details for
Quantity
  • Availability
  • Add to Cart