RIE156330125 Rieke Metals, Inc.

2,2-Dimethyl-4-oxo-4-(4-n-pentylphenyl)butyric acid

  • MDL number MFCD09801839
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7273

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7273
Availability details for
Quantity
  • Availability
  • Add to Cart