RIE156330126 Rieke Metals, Inc.

3-Methyl-5-oxo-5-(4-n-pentylphenyl)valeric acid

  • MDL number MFCD09801840
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7274

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7274
Availability details for
Quantity
  • Availability
  • Add to Cart