RIE156330135 Rieke Metals, Inc.

2,2-Dimethyl-4-oxo-4-(2,3,4,5,6-pentamethylphenyl)butyric acid

  • MDL number MFCD09801849
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7283

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7283
Availability details for
Quantity
  • Availability
  • Add to Cart