RIE156330138 Rieke Metals, Inc.

5-(3,5-Dimethyl-4-methoxyphenyl)-3-methyl-5-oxovaleric acid

  • MDL number MFCD09801852
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7286

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7286
Availability details for
Quantity
  • Availability
  • Add to Cart